Benzyltriethylammonium tetrafluoroborate 98+%

Supplier: Ambeed
Warning

Synonyms: N-Benzyl-N,N-diethylethanaminium tetrafluoroborate

A631140-100G A631140-25G A631140-5G
76696-788EA 174 USD
76696-788 76696-790 76696-792
Benzyltriethylammonium tetrafluoroborate 98+%
Benzyltriethylammonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₃H₂₂BF₄N
MW: 279.13 g/mol
MDL Number: MFCD00011823
CAS Number: 77794-93-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR