Butylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Butan-1-amine hydrochloride, 1-Aminobutane Hydrochloride

A383644-100G A383644-1G A383644-25G A383644-5G
76694-520EA 54 USD
76694-520 76694-522 76694-524 76694-526
Butylamine hydrochloride 98%
Butylamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁N
MW: 109.60 g/mol
MDL Number: MFCD00043791
CAS Number: 3858-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR