3-(Bromomethyl)-2-chloro-6-fluoropyridine 97%

Supplier: Ambeed
Danger

A1205516-250MG
76677-594EA 381 USD
76677-594
3-(Bromomethyl)-2-chloro-6-fluoropyridine 97%
3-(Bromomethyl)-2-chloro-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MW: 224.46 g/mol
MDL Number: MFCD16607615
CAS Number: 1227516-84-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR