2-Chloro-4-methylthiazole 98+%

Supplier: Ambeed
Warning

A1490710-1G
76665-832EA 18 USD
76665-832
2-Chloro-4-methylthiazole 98+%
2-Chloro-4-methylthiazole

Precaución: for Research Use Only

Formula: C₄H₄ClNS
MW: 133.599 g/mol
MDL Number: MFCD00051033
CAS Number: 26847-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR