(3-Chloro-5-fluorobenzyl)hydrazine hydrochloride 97%

Supplier: Ambeed
Danger

A1352256-100MG A1352256-1G A1352256-250MG
76658-536EA 26 USD
76658-536 76658-538 76658-540
(3-Chloro-5-fluorobenzyl)hydrazine hydrochloride 97%
(3-Chloro-5-fluorobenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂FN₂
MW: 211.0642 g/mol
MDL Number: MFCD31559418
CAS Number: 2137655-30-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR