5-Bromo-2-methoxythioanisole 95%

Supplier: Ambeed
Warning

A714198-250MG
76651-424EA 60.48 USD
76651-424
5-Bromo-2-methoxythioanisole 95%
5-Bromo-2-methoxythioanisole

Precaución: for Research Use Only

Formula: C₈H₉BrOS
MW: 233.1255 g/mol
MDL Number: MFCD22488925
CAS Number: 66623-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR