N1-Methylbutane-1,4-diamine dihydrochloride 95%

Supplier: Ambeed
Warning

A704007-250MG
76651-244EA 318 USD
76651-244
N1-Methylbutane-1,4-diamine dihydrochloride 95%
N1-Methylbutane-1,4-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₆Cl₂N₂
MW: 175.0999 g/mol
MDL Number: MFCD00053031
CAS Number: 89690-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR