Ethyl 1-(aminomethyl)cyclobutanecarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A676767-100MG A676767-1G A676767-250MG
76650-916EA 49 USD
76650-916 76650-918 76650-920
Ethyl 1-(aminomethyl)cyclobutanecarboxylate hydrochloride 98%
Ethyl 1-(aminomethyl)cyclobutanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₂
MW: 193.6711 g/mol
MDL Number: MFCD18483101
CAS Number: 1269021-92-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR