Capillary Arrays
Supplier: Agilent Technologies
A2300-1250-2247 M5310AA-001 M5310AA-002 M5310AA-003 M5311AA-001 M5311AA-002 M5311AA-003 A2300-1250-3355 A2300-1250-5580 A2300-4850-3355 A2300-9650-3355 A2300-9650-5580 M5312AA-001 M5312AA-002 M5320AA-001 M5320AA-002
76645-418EA 1391.05 USD
76645-418 76645-078 76645-064 76645-092 76645-440 76645-178 76645-168 76645-074 76645-386 76645-234 76645-462 76645-208 76645-312 76645-198 76645-404 76645-144
Capillary Arrays
Electrophoresis Systems Capillary Electrophoresis Systems
Capillary arrays for fragment analyzer and ZAG systems.
  • Array cartridges are robust and have a long lifespan, keeping your instrument on-task from one project to the next
  • Short arrays provide robust separation for most samples
  • Speed up separation times and increase overall QC throughput with ultra-short arrays
  • Enhance separation resolution and focus nucleic acid analysis with long arrays
The fragment analyzer systems offer a selection of capillary arrays to optimize your QC workflow by separating nucleic acids via parallel capillary electrophoresis.More Product Information