Bioanalyzer High Sensitivity Protein Analysis
Supplier: Agilent Technologies
5067-1575 5067-1576 5067-1577 5067-1578
76645-002EA 1370.63 USD
76645-002 76645-030 76645-230 76645-430
Bioanalyzer High Sensitivity Protein Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
High sensitivity electrophoretic separation of proteins and peptides up to 250 kD.
  • A ready-to-use system featuring standardized assays and pre-packaged reagent kits which are suitable to analyze samples up to 250 kD
  • After sample labeling, fast results are achieved with automated electrophoresis of 10 samples in 30 minutes
  • Highly sensitive with an accuracy down to silver stain levels, approximately 1 pg/µl of labeled protein on chip
  • Fully operational across a large linear dynamic range with four orders of magnitude
  • Highly precise impurity detection, down to
More Product Information