VWR® Performance Flammable Material Storage Manual Defrost Freezers with Inner Doors
Supplier: VWR International
76644-708 76644-712 76644-716
76644-708EA 9198.07 USD
76644-708 76644-712 76644-716
VWR® Performance Flammable Material Storage Manual Defrost Freezers with Inner Doors
Freezers Flammable Material Freezers
The inner doors on these flammable storage freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials inMore Product Information