VWR® Heavy-Duty Absorbent Wipers
Supplier: VWR International
AVTR1 AVTR2 AVTR3
76639-024CS 160.85 USD
76639-024 76639-030 76639-026
VWR® Heavy-Duty Absorbent Wipers
Wipes Disposable Wipes and Towels Wipes for General Applications
VWR® Industrial absorbent wipers are used in many industries and are made from recycled materials. These wipers are designed to be durable and resistant to abrasion.
  • Durable and absorbent heavy-duty wipers
  • Lint-free
  • Strong either wet or dry
  • Flat, bulk in box
These top‑tier wipers, renowned for their exceptional cleanliness and absorbency, are designed to excel in demanding environments. Crafted with a high‑performance fiber blend, they boast unparalleled lint control, ensuring a flawless finish with minimal risk of contamination. Engineered to resistMore Product Information