Individual PCR Tubes with Attached Caps, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0790 4TI-0790/2D 4TI-0795
76638-066CS 78.14 USD
76638-066 76638-082 76638-210
Individual PCR Tubes with Attached Caps, Azenta Life Sciences
Tubes PCR Tubes
High profile, 0.2 ml wells, clear polypropylene; working volume: <200 µl, total well capacity: 300 µl.
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • High profile: <0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity
  • Snap-shut cap design
  • Recommended for low to medium throughput applications
  • Our individual tubes with flat caps are also available with a unique 2D code applied to the top of each tube
  • This allows for
More Product Information