PCR Plates, 96-Well, Individual Access, Optical Bottom, Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0970/RA 4TI-0970/RA-F
76638-158CS 379.51 USD
76638-158 76638-188
PCR Plates, 96-Well, Individual Access, Optical Bottom, Skirted, Azenta Life Sciences
Microplates PCR Plates
These plates are ideal for use with robotic systems. Low profile, flat optical bottom with individually removable 0.1 ml polypropylene wells, rigid polycarbonate frame, cut corner H1.
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile <0.1 ml (100 µl) working volume, <0.18ml (180 µl) total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Rigid
More Product Information