PCR Plate Frames for Removable 8-Well Tube Strips, 96-Well, Fully Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0753/757 4TI-0753/950W 4TI-0753/C/757 4TI-0757-F 4TI-0950/W-F
76637-982CS 769.84 USD
76637-982 76638-046 76638-150 76638-016 76629-104
PCR Plate Frames for Removable 8-Well Tube Strips, 96-Well, Fully Skirted, Azenta Life Sciences
Microplates PCR Plates
Rigid polycarbonate frame, for use with removable 8 well tube strips available for use on Roche LightCycler® 480 and for universal use.
  • Free from DNase, RNase, human genomic DNA,and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well
More Product Information