PCR Tube Strips, 8-Well, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0753 4TI-0753/C 4TI-0754 4TI-0754/C
76638-070CS 124.94 USD
76638-070 76638-128 76637-996 76638-062
PCR Tube Strips, 8-Well, Azenta Life Sciences
Tubes PCR Tubes
Strips of 8 polypropylene PCR tubes. Can be used alone or in conjunction with any 96-well PCR plate frame.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile: <0.1 ml (100 µl) working volume, 0.2 ml (200 µl) total well capacity when used with sealing options
  • Can be used as individual strips, or in a 96 Well PCR Plate frame for easier handling
  • Compatible with standard multichannel pipettes
These low profile PCR tube stripsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable