8-Well PCR Tube Strips, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0780 4TI-0781 4TI-0784
76637-958CS 164.38 USD
76637-958 76637-968 76638-030
8-Well PCR Tube Strips, Azenta Life Sciences
Tubes PCR Tubes
High profile, 0.2 ml wells, clear polypropylene; working volume: <200 µl, total well capacity: 300 µl.
  • Suitable for all standard 0.2 ml block thermal cyclers
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • High profile: <0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity when used with sealing options
  • Individually numbered tubes
  • Can be cut into sections
These PCR tubes are molded from virgin polypropylene in aMore Product Information