SUPERFLO™ Histology Cassettes, One-Piece Molded, Simport Scientific
Supplier: Simport Scientific
M530-10 M530-11 M530-12 M530-2 M531-2 M531-3 M531-4 M531-5 M531-6 M531-7 M531-8 M531-9 M532-10 M532-11 M530-3 M530-4 M530-5 M530-6 M530-7 M530-8 M530-9 M531-10 M531-11 M531-12 M532-12 M532-2 M532-3 M532-4 M532-5 M532-6 M532-7 M532-8 M532-9 M430-10T M430-11T M430-12T M430-2T M430-3T M430-4T M430-5T M430-6T M430-7T M430-8T M431-8T M431-9T M432-10T M432-11T M432-12T M432-2T M432-3T M432-4T M432-5T M432-6T M432-7T M432-8T M432-9T M430-9T M431-10T M431-11T M431-12T M431-2T M431-3T M431-4T M431-5T M431-6T M431-7T
76636-102CS 221.87 USD
76636-102 76636-104 76636-106 76636-086 76636-108 76636-110 76636-112 76636-114 76636-116 76636-118 76636-120 76636-122 76636-146 76636-148 76636-088 76636-090 76636-092 76636-094 76636-096 76636-098 76636-100 76636-124 76636-126 76636-128 76636-150 76636-130 76636-132 76636-134 76636-136 76636-138 76636-140 76636-142 76636-144 76635-970 76635-972 76635-974 76635-954 76635-956 76635-958 76635-960 76635-962 76635-964 76635-966 76635-988 76635-990 76636-014 76636-016 76636-018 76635-998 76636-000 76636-002 76636-004 76636-006 76636-008 76636-010 76636-012 76635-968 76635-992 76635-994 76635-996 76635-976 76635-978 76635-980 76635-982 76635-984 76635-986
SUPERFLO™ Histology Cassettes, One-Piece Molded, Simport Scientific
Embedding Cassettes Tissue Processing Cassettes
This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss.
  • Unique lateral vents, fluids can access tissue from all four sides
  • Basket style for small tissue management
  • One-piece molded with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colors
Molded from high-density polymer, it keeps specimens safely submerged in liquid.
Available for tissue, biopsy and micro-More Product Information