Paraffin wax, POLYFin™
Supplier: General Data Healthcare
Synonyms: Wax Paraffin, Paraffin wax
H-PF
76635-704CS 259.46 USD
76635-704
Paraffin wax, POLYFin™
Paraffin
POLYFin is the preferred histology paraffin wax that delivers stability in handling, excellent tissue infiltration and support for precise and efficient tissue cutting at the microtome station.POLYFin is ideal for labs that require a consistent paraffin formulation for their wax blocks that is dependable through multiple shifts.
  • POLYFin™ delivers a consistent and dependable formulation with a lower melting point for efficient tissue embedding, processing and cutting
  • POLYFin is ideal for histotechs who require efficient, precise cutting of paraffin wax blocks at the microtome station with no errors
CAS Number: 8002-74-2
Order Now
SPECIFICATIONS
Environmentally Preferable