FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0770/C 4TI-0770 4TI-OX770/C/SBC 4TI-LB0770/C 4TI-0770/B 4TI-0770/C/10P 4TI-0770/G 4TI-0770/R 4TI-0770/X 4TI-0771 4TI-0772 4TI-0772/10P
76619-070CS 307.46 USD
76619-070 76629-154 76638-022 76629-146 76629-158 76629-052 76629-162 76629-064 76629-108 76629-004 76629-094 76629-134
FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences
Microplates PCR Plates
High profile, 0.2 ml polypropylene wells, polycarbonate frame, cut corner A12, working volume: <200 µl, total well capacity: 300 µl. Ideal for use with robotic systems. Ideal for ABI® thermal cyclers and sequencers. Compatible with standard multichannel pipettes.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample
More Product Information