PCR Tube Strips, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0785/B 4TI-0785/G 4TI-0785/M 4TI-0785/P 4TI-0785/R 4TI-0785/W/2D 4TI-0785/X 4TI-0785/X/2D 4TI-0785/XW 4TI-0786/B 4TI-0786/G 4TI-0786/M 4TI-0786/P 4TI-0786/R 4TI-0786/W/2D 4TI-0786/X 4TI-0786/X/2D 4TI-0786/XW
76629-026CS 161.41 USD
76629-026 76629-016 76629-062 76629-156 76629-164 76629-124 76629-068 76629-044 76629-056 76629-140 76629-112 76629-024 76629-070 76629-074 76629-130 76629-022 76629-120 76629-092
PCR Tube Strips, Azenta Life Sciences
Microplates PCR Plates
Strips of eight polypropylene PCR tubes held in a rigid polycarbonate frame for highest mechanical stability.
  • Two component design in an eight well strip format
  • Compatible with the majority of thermal cyclers
  • Available with either domed cap strips or with flat, optically clear cap strips
  • Available with off-the-shelf 2D code
By molding the frame portion in a more rigid polymer, the mechanical stability is greatly improved compared with traditional single piece products.
For color coding of experiments, we offer strips with clear wells in sixMore Product Information