1-​azido-​9-​bromononane 95%

Supplier: AFG BIOSCIENCE LLC

C56774-1G C56774-2G C56774-500MG
76620-306EA 739.39 USD
76620-306 76620-308 76620-304
1-​azido-​9-​bromononane 95%
1-​azido-​9-​bromononane
Formula: C₉H₁₈BrN₃
MW: 248.16 g/mol
Storage Temperature: Refrigerator
CAS Number: 1262683-54-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance Colorless oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR