3-Azido-3-deoxy-D-glucopyranose 95

Supplier: AFG BIOSCIENCE LLC

C51784-1G C51784-500MG C51784-250MG
76619-698EA 443.63 USD
76619-698 76619-696 77888-818
3-Azido-3-deoxy-D-glucopyranose 95
3-Azido-3-deoxy-D-glucopyranose
Formula: C₆H₁₁N₃O₅
MW: 205.17 g/mol
Storage Temperature: Freezer
CAS Number: 104875-44-7
New Product

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Colorless Low-Melting Solid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR