Klercide™ Extendable Handle for Mop Frames, Stainless Steel
Supplier: ECOLAB LIFE SCIENCES
10056565
76554-218EA 919.02 USD
76554-218
Klercide™ Extendable Handle for Mop Frames, Stainless Steel
Mops
Ecolab's stainless steel extendable handle for mop frames is a lightweight extendable handle (up to 6.89 ft) that enables high walls, ceilings, and other raised platforms to be easily reached.
The extendable handle can easily be dismantled for complete sterilization by autoclave.