Antibac B, Highly-soluble Chlorinated Sanitizer
Supplier: DIVERSEY INC.
08514210
76547-720EA 2959.99 USD
76547-720
Antibac B, Highly-soluble Chlorinated Sanitizer
Cleaning Agents Detergents and Surface Cleansers
Antibac B sanitizer is a rapid acting, highly soluble, granular, organic chlorinated solution.
  • Readily soluble for preparing stock solutions used in automatic sanitizing units.
  • Leaves no films on equiptment.
  • Can be used at 100 ppm for most sanitizing operations.
It is designed for use as a hard surface contact sanitizer in dairy plants, food processing plants, and pharmaceutical and cosmetics manufacturing facilities.
It can also be used for sanitizing commercial eggs.