Soil Hydrometer, Specific Gravity, ASTM 151H
Supplier: Heathrow Scientific
67151H
76546-590EA 50.34 USD
76546-590
Soil Hydrometer, Specific Gravity, ASTM 151H
Hydrometers
This hydrometer measures the specific gravity of soil samples in suspension.
  • Calibrated for use at 20 °C (68 °F)
This hydrometer is often used with a sedimentation cylinder for soil analysis.
Certificaciones: Manufactured to the specifications of the American Society for Testing and Materials (ASTM).