VWR® Inoculating Loops and Needles, Sterile
Supplier: VWR International
76544-924 76544-926 76544-928 76544-930 76544-932 76544-934 76544-936 76544-938 76544-940
76544-924BX 81.88 USD
76544-924 76544-926 76544-928 76544-930 76544-932 76544-934 76544-936 76544-938 76544-940
VWR® Inoculating Loops and Needles, Sterile
Spreaders Inoculating Loops and Needles
High-impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface.
  • Polystyrene fabrication
  • Smooth and flexible
  • Sterile
Needles are straight and suitable for removal of single colony specimens. Packed sterile in safe, tamperproof, zip-seal resealable bags. Disposable inoculating loops and needles do not need flaming, eliminating the risk of infection due to aerosol formation of pathogenic substances as well as cross-contamination due to improper sterilization. Loops and needlesMore Product Information