Methylone (hydrochloride) (CRM), Cayman Chemical

Supplier: Cayman Chemical Company

20891-1
76544-224EA 182 USD
76544-224
Methylone (hydrochloride) (CRM), Cayman Chemical
Standards Organic Standards

A Certified Reference Material


  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard

Order Now

SPECIFICATIONS

Description Methylone (hydrochloride)
Synonym(s) M1; bk-MDMA; 3,4-Methylenedioxy-N-methylcathinone; β-keto Methylenedioxy Methamphetamine
Environmentally Preferable
CAS No. 186028-80-8
Formula C₁₁H₁₃NO₃·HCl
Storage Temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR