VWR® Scintillation Vials with Screw Cap, Borosilicate Glass
Supplier: VWR International
086.208.01 086.210.01 086.210.02 086.210.03 086.209.01 086.208.02 086.208.03 086.208.04
76538-954CS 217.29 USD
76538-954 76538-956 76538-958 76538-960 76538-962 76538-964 76538-966 76538-968
VWR® Scintillation Vials with Screw Cap, Borosilicate Glass
Vials
VWR® scintillation vials are produced from borosilicate 3.3 glass. They are compatible with all scintillation counters.
  • Borosilicate glass construction
  • Fit all standard liquid scintillation counting equipment
  • High resistance against chemicals and thermal shock
  • Low background counts and high ultraviolet transmission
  • Choose attached or unattached caps
VWR® scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting or for sample storage.
Each vials includes a white screw cap with a top suitable for marking.
Order Now
SPECIFICATIONS
Environmentally Preferable