Melting Furnaces, Nabertherm
Supplier: NABERTHERM INC.
KD0110IRNA-208 KD0110IRNA-240 KD0130IRNA-208 KD0130IRNA-240 KD0210IRNA-208 KD0210IRNA-240 KD0230IRNA-208 KD0230IRNA-240 KD0410IRNA-208 KD0410IRNA-240 KD0430IRNA-480 KC0143IENC-480 KC0243IENC-480
76538-294EA 8647.63 USD
76538-294 76538-296 76538-298 76538-300 76538-302 76538-304 76538-306 76538-308 76538-310 76538-312 76538-314 76538-316 76538-318
Melting Furnaces, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These compact melting furnaces for the melting of non-ferrous metals and alloys are one of a kind and have a number of technical advantages
  • Tmax 1000, 1300 or 1500 °C, with melt temperature about 80 to 110 °C lower
  • Crucible sizes of 0.75, 1.5 or 3 liters
  • Crucible with integrated pouring spout of iso-graphite included with delivery
  • Additional spout (not for KC), mounted at the furnace for exact pouring
  • Compact bench-top design, simple emptying of crucible by tiltiing system with gas damper
  • Crucible for heating up of melting furnace insulated with a hinged
More Product Information