Muffle Furnaces with Scale and Software, Tmax 1200 °C, L 9/12 SW or LT 9/12 SW, Nabertherm
Supplier: NABERTHERM INC.
LW094K2LN2-208 LW094K2LN2-240 LW094K2ON2-208 LW094K2ON2-240 LW094H2LN2-208 LW094H2LN2-240 LW094H2ON2-208 LW094H2ON2-240
76538-174EA 19944.3 USD
76538-174 76538-176 76538-178 76538-180 76538-190 76538-192 76538-194 76538-196
Muffle Furnaces with Scale and Software, Tmax 1200 °C, L 9/12 SW or LT 9/12 SW, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These weighing furnaces with integrated precision scale and software, were designed especially for determining combustion loss in the laboratory. The determination of combustion loss is necessary, for instance, when analyzing sludges. Combustion loss is defined as the difference between the initial total mass and the combustion residue. During the process the software supplied records both temperature and weight loss.
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and easy to replace
More Product Information