Concept Lab Benches
Supplier: Treston
10149031 10149032 10149033 10549031 10549032 10549033 14-C10049121 14-C10049403 14-C10049122 14-C10049123 14-C10049124 14-C10049125 14-C10149134 14-C10149403 14-C10149132 14-C10149123 14-C10149124 14-C10149125 14-C10349133 14-C10349403 14-C10349132 14-C10349123 14-C10349124 14-C10349125 14-C10049440 14-C10049441 14-C10049442 14-C10049443 14-C10049444 14-C10049445 14-C10149440 14-C10149441 14-C10149442 14-C10149443 14-C10149444 14-C10149445 14-C10349440 14-C10349441 14-C10349442 14-C10349443 14-C10349444 14-C10349445 14-C10049393 14-C10049394 14-C10049365 14-C10049404 14-C10049405 14-C10049406 14-C10149393 14-C10149394 14-C10149395 14-C10149396 14-C10149397 14-C10149398 14-C10349393 14-C10349394 14-C10349395 14-C10349396 14-C10349397 14-C10349398 14-C10049446 14-C10049447 14-C10049448 14-C10049449 14-C10049450 14-C10049451 14-C10149446 14-C10149447 14-C10149448 14-C10149449 14-C10149450 14-C10149451 14-C10349446 14-C10349447 14-C10349448 14-C10349449 14-C10349450 14-C10349451 14-C10049420 14-C10049421 14-C10049422 14-C10049423 14-C10049424 14-C10049425 14-C10149420 14-C10149421 14-C10149422 14-C10149423 14-C10149424 14-C10149425 14-C10349420 14-C10349421 14-C10349422 14-C10349423 14-C10349424 14-C10349425 14-C10049456 14-C10049457 14-C10049458 14-C10049459 14-C10049460 14-C10049461 14-C10149456 14-C10149457 14-C10149458 14-C10149459 14-C10149460 14-C10149461 14-C10349456 14-C10349457 14-C10349458 14-C10349459 14-C10349460 14-C10349461 14-C10049426 14-C10049427 14-C10049428 14-C10049429 14-C10049430 14-C10049431 14-C10149426 14-C10149427 14-C10149428 14-C10149429 14-C10149430 14-C10149431 14-C10349426 14-C10349427 14-C10349428 14-C10349429 14-C10349430 14-C10349431 14-C10049462 14-C10049463 14-C10049464 14-C10049465 14-C10049466 14-C10049467 14-C10149462 14-C10149463 14-C10149464 14-C10149465 14-C10149466 14-C10149467 14-C10349462 14-C10349463 14-C10349464 14-C10349465 14-C10349466 14-C10349467 10049031P 10049032P 10049033P
89202-092EA 3258.3 USD
89202-092 89202-094 89202-096 89202-098 89202-100 89202-102 76532-606 76532-608 76532-610 76532-612 76532-614 76532-616 76532-618 76532-620 76532-622 76532-624 76532-626 76532-628 76532-630 76532-632 76532-634 76532-636 76532-638 76532-640 76532-642 76532-644 76532-646 76532-648 76532-650 76532-652 76532-654 76532-656 76532-658 76532-660 76532-662 76532-664 76532-666 76532-668 76532-670 76532-672 76532-674 76532-676 76532-678 76532-680 76532-682 76532-684 76532-686 76532-688 76532-690 76532-692 76532-694 76532-696 76532-698 76532-700 76532-702 76532-704 76532-706 76532-708 76532-710 76532-712 76532-714 76532-716 76532-718 76532-720 76532-722 76532-724 76532-726 76532-728 76532-730 76532-732 76532-734 76532-736 76532-738 76532-740 76532-742 76532-744 76532-746 76532-748 76532-750 76532-752 76532-754 76532-756 76532-758 76532-760 76532-762 76532-764 76532-766 76532-768 76532-770 76532-772 76532-774 76532-776 76532-778 76532-780 76532-782 76532-784 76532-786 76532-788 76532-790 76532-792 76532-794 76532-796 76532-798 76532-800 76532-802 76532-804 76532-806 76532-808 76532-810 76532-812 76532-814 76532-816 76532-818 76532-820 76532-822 76532-824 76532-826 76532-828 76532-830 76532-832 76532-834 76532-836 76532-838 76532-840 76532-842 76532-844 76532-846 76532-848 76532-850 76532-852 76532-854 76532-856 76532-858 76532-860 76532-862 76532-864 76532-866 76532-868 76532-870 76532-872 76532-874 76532-876 76532-878 76532-880 76532-882 76532-884 76532-886 76532-888 76532-890 76532-892 76533-018 76533-020 76533-022
Concept Lab Benches
Furniture Tables and Workbenches
Treston® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Powder coated steel
  • Suitable for heavy loads up to 1100 lbs. (depending on model)
  • Wide range of accessories available to meet any need
  • QuickShip available
  • 10-Year Warranty
  • Paint color: Light gray
Treston® lab benches are ergonomically designed to meet the needs of laboratory personnel. There areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable