VWR®, Duster Spray
Supplier: VWR International
76528-070
76528-070EA 48.93 USD
76528-070
VWR®, Duster Spray
Sprays
Easily remove debris from hard-to-reach places.
  • All-purpose sterilized cleaner is nonflammable
  • Remove dust, dirt, and microscopic debris from home and office equipment
  • Safe for the ozone with ultra-low global warming potential (GWP)
VWR® air duster spray removes dust, dirt, and debris from delicate or hard-to-reach places such as keyboards, copiers, lab equipment, fans, and more. The aerosol cleaning spray provides jets of cleaning power that is easy and safe to use.
Información de suministro: Extension tube.