TeenPrep™ Adapter, 12×15 ml, MP Biomedicals
Supplier: MP Biomedicals
116002526
76522-162EA 1759.93 USD
76522-162
TeenPrep™ Adapter, 12×15 ml, MP Biomedicals
Homogenizers Benchtop Homogenizers
Adapter for FastPrep-24 instruments.
  • 12×15 ml tubes
  • For room temperature lysis and grinding of samples
Medium-size adapter enables FastPrep-24 instruments to accommodate 12×15 ml Lysing Matrix tubes to ensure thorough homogenization of any sample type in seconds.
Interchangeable adapters for FastPrep instruments accommodate various Lysing Matrix tube sizes for sample processing. The FastPrep bead beating system maximizes mechanical tissue homogenization for efficient isolation of high-quality DNA, RNA, and proteins, without the need for chemicalsMore Product Information