Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Supplier: Horizon Scientific
PRF102WWW/030 PRF102WWW/0 PRF102WWG/040 PRF102WWG/030 PRF102WWG/0
76516-560EA 4432.81 USD
76516-560 76516-562 76516-564 76516-566 76516-568
Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Refrigerators
These efficient refrigerator and freezer combo units provide quality cold storage in laboratory settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Natural, hydrocarbon refrigerant
  • White cabinet and door
These White Diamond combo units offer a cold-storage solution for a variety of laboratory needs. Units feature a series of temperature alarms to keep stored products at peak effectiveness. All models are equipped withMore Product Information