ABS® Vaccine Refrigerators, Certified to the NSF/ANSI 456 Standard, Horizon Scientific
Supplier: Horizon Scientific
PH-ABT-NSF-23G PH-ABT-NSF-S23G PH-ABT-NSF-23S PH-ABT-NSF-S23S
76516-538EA 10472.48 USD
76516-538 76516-540 76516-542 76516-544
ABS® Vaccine Refrigerators, Certified to the NSF/ANSI 456 Standard, Horizon Scientific
Refrigerators
These vaccine refrigerators are certified to the NSF/ANSI 456 Standard for Vaccine Storage.
  • Microprocessor temperature controller
  • High / low temperature alarms
  • Probe access port (³/₄")
  • LED interior lighting
  • Natural, hydrocarbon refrigerant
  • Pharmacy toolkit and temperature logs
  • Remote alarm contacts
  • ENERGY STAR certified*
These units protect stored contents with greater stringency as outlined by the NSF/ANSI 456 Standard for Vaccine Storage. Units feature superior temperature stability and uniformity, quick recovery times after door openings, andMore Product Information