OS20L Digital Overhead Stirrers, Benchmark Scientific
Supplier: VWR International
76498-684
76498-684EA 977.08 USD
76498-684
OS20L Digital Overhead Stirrers, Benchmark Scientific
Stirrers Overhead Stirrers
High torque and advanced digital microprocessor control for smooth mixing of samples up to 20 L.
  • LCD control panel
  • Digital accuracy and convenience
  • Compact and quiet
  • Includes complete stand assembly
The OS20L overhead stirrer is suitable for mixing low to medium viscosity samples up to 20 L. Speed and run time are accurately set and monitored using the digital LCD screen. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools.
The overhead stirrer is protected from damage by sensors that shutMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable