Chromaster HPLC System, Hitachi
Supplier: HITACHI HIGH-TECH AMERICA, INC.
889-0136 892-0106 892-0116 892-0126 892-0136 892-0226 892-0286 892-0306 892-0406 892-0426 892-0446 892-0486 892-0606 892-0787 892-0797 892-0806
76493-026EA 7911.31 USD
76493-026 76493-058 76493-060 76493-062 76493-064 76493-092 76493-128 76493-132 76493-142 76493-144 76493-146 76493-148 76493-174 76493-176 76493-178 76493-180
Chromaster HPLC System, Hitachi
Chromatography Systems Complete HPLC Systems
The Chromaster HPLC system has established new limits in performance, ease of use, and robustness. Its design incorporates an extremely low dwell volume and a very low carry-over. In combination with very sensitive detector technology, especially the fluorescence detector, the Chromaster HPLC system is one of the strongest LC systems on the market. A huge amount of accessories and a modular set-up, enable an easy adaption to all customer needs, whether if it is used for an aggressive industrial or a high sensitive pharma application. VWR's relationship with Hitachi started over 30 years agoMore Product Information