VWR® Calibrated Digital Verification Thermometers, General Use
Supplier: VWR International
76491-658 76491-660 76491-662
76491-658CS 553.2 USD
76491-658 76491-660 76491-662
VWR® Calibrated Digital Verification Thermometers, General Use
Thermometers Digital Thermometers
Digital thermometers verify sample temperatures in freezers, refrigerators, and incubators.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in the USA, traceable to NIST
Available calibrated for specific applications and equipment.
Certificaciones: ISO 17025, ISO/IEC 17025:2005, traceable to NIST.
Información de suministro: Includes multi-More Product Information