OHM Metrics™ Digital Display ESD Tester and Footplate, Transforming Technologies
Supplier: Transforming Technologies
PDT800
76490-158EA 2077.38 USD
76490-158
OHM Metrics™ Digital Display ESD Tester and Footplate, Transforming Technologies
Static Control Monitors
This OHM Metrics™ unit is a comprehensive, accurate ESD ground device tester that fully meets daily ESD checking needs.
  • Tests wrist straps and footwear
  • Proactive 'near-fail' alert indicates if a ground device is close to fail levels
  • Eliminate failures in the field
  • Preset with broad selectable test range: 100 KΩ to 1 GΩ
An easy, accurate, and time saving one-touch digital wrist strap and footwear tester. Simultaneously and independently test both feet and a wrist strap while filtering out human body noise. The results are visible via a digital screen.More Product Information