Wolfgang Warmbier Backrest Probe, Transforming Technologies
Supplier: Transforming Technologies
7220.900
76489-574EA 1940.68 USD
76489-574
Wolfgang Warmbier Backrest Probe, Transforming Technologies
Static Control Monitors
The Backrest probe is designed to evaluate ESD chair backrests according to ANSI ESD20.20.
  • Ensures excellent contact during evaluations of ESD chairs
  • Rugged metal probe
  • 4 mm banana plug socket connection
For use with resistance meters.