SP Bel-Art H-B Durac® Calibrated Thermocouple Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B60901-0800 B60901-1000 B60901-1100 B60911-0200
76486-046CS 850.35 USD
76486-046 76486-048 76486-050 76486-052
SP Bel-Art H-B Durac® Calibrated Thermocouple Thermometers, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Used for temperature measurement in life science research, kilns, gas turbine exhaust, diesel engines, and other industrial processes, H-B Calibrated Thermocouple Thermometers provide measurement throughout a broad temperature range and the ability to utilize multiple probes at one time.
  • Wide temperature range allows accurate monitoring at extreme temperatures
  • Max/min/average temperature recording capability
  • Data hold
  • Software CD (where included) for real time data collection and logging via USB
  • Bluetooth® app available for use with B60911-0200