Cooled Incubators with Advanced Peltier Technology, IPP and IPPeco Series, Memmert
Supplier: Memmert
IPP110ECO IPP110ECOPLUS IPP260ECOPLUS IPP260ECO IPP410ECO IPP410ECOPLUS IPP750ECO IPP750ECOPLUS IPP2200ECO IPP2200ECO+ IPP1060ECO IPP1060ECO+ IPP1400ECO IPP1400ECO+
76485-924EA 7885.04 USD
76485-924 76485-926 76485-928 76485-930 76485-932 76485-934 76485-936 76485-938 76632-922 76632-924 76632-914 76632-916 76632-918 76632-920
Cooled Incubators with Advanced Peltier Technology, IPP and IPPeco Series, Memmert
Incubators Cooled Incubators
Memmert’s Cooled Storage Incubator (IPPeco) is the most energy-efficient and powerful cooled incubator of its kind due to state-of-the-art Peltier technology. Heat-up, cool down and recovery times are much shorter than comparable incubators. The Peltier-cooled incubator from Memmert saves on operating costs, ensures sample uniformity and makes work more efficient. 
  • From temps of at least 20°C below ambient up to +70°C
  • Energy saving Peltier heating/cooling system integrated in the rear
  • Programmable with internal data logging via included AtmoCONTROL
More Product Information