iFluor® 488-dUTP 1 mM in TRIS buffer (pH 7.5)

Supplier: AAT BIOQUEST INC
Warning

17039
76485-814EA 112.97 USD
76485-814
iFluor® 488-dUTP 1 mM in TRIS buffer (pH 7.5)
iFluor® 488-dUTP

The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • Useful for DNA probe development
  • Well-cited
  • Multiple applications


Precaución: For research use only

MW: 1152.83 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR