5-dROX succinimidyl ester
Supplier: AAT BIOQUEST INC
312
76485-392EA 2342.12 USD
76485-392
5-dROX succinimidyl ester
5-dROX succinimidyl ester
5-dROX (5-Carboxy-4,7-dichloror-X-hodamine) is one of the four critical 4,7-dichloro-rhodamine dyes used as the BigDye terminators for DNA sequencing in combination with dR110, dR6G and dROX.
  • Sanger sequencing building block
Precaución: For Research Use Only