phosphatase reaction stopping solution, Signal Guard™
Supplier: AAT BIOQUEST INC
Warning
11622
76484-828EA 124.31 USD
76484-828
phosphatase reaction stopping solution, Signal Guard™
phosphatase reaction stopping solution
Alkaline and acid phosphatases are the hydrolase enzymes responsible for dephosphorylation of molecules such as nucleotides, proteins and alkaloids under alkaline and acidic conditions, respectively.
  • compatible with all major phosphatase groups
  • neutral and mild
  • ready to use
  • stabilizes signal for up to 18 hours
  • compatible with colorimetric and fluorimetric substrates
ADR: 5 II