Cal-770™ potassium salt

Supplier: AAT BIOQUEST INC
Warning

20460
76484-646EA 500.94 USD
76484-646
Cal-770™ potassium salt
Cal-770™ potassium salt

Calcium measurement is critical for numerous biological investigations.


  • Higher sensitivity
  • Specific to intracellular calcium
  • Steady signal
  • Cell impermeant


Precaución: For research use only

MW: 1688.58 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR