Amplite® Colorimetric Sphingomyelinase Assay Kit *Blue Color* , AAT Bioquest
Supplier: AAT BIOQUEST INC
13620
76482-212EA 249.58 USD
76482-212
Amplite® Colorimetric Sphingomyelinase Assay Kit *Blue Color* , AAT Bioquest
Assays Enzyme Assays
Sphingomyelinase (SMase) is an enzyme that is responsible for cleaving sphingomyelin (SM) to phosphocholine and ceramide.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Precaución: For research use only.