Hydrogen peroxide solution 50 mM stabilised, ReadiUse™

Supplier: AAT BIOQUEST INC
Warning

11004
76481-876EA 124.31 USD
76481-876
Hydrogen peroxide solution 50 mM stabilised, ReadiUse™
Hydrogen peroxide

Compared to other commercial hydrogen peroxide solutions, our formulated hydrogen peroxide solution is much more stable.


  • improved stability
  • increases assay reproducibility

Formula: H₂O₂
MW: 34.01
Boiling Pt: 150.2
Melting Pt: -0.41
Density: 1.11
MDL Number: MFCD00011333
CAS Number: 7722-84-1
ADR: 5 II
Merck Index: 14,04798

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR