ReadiLink™ Rapid iFluor® 594 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Supplier: AAT BIOQUEST INC
1230
76481-258EA 188.87 USD
76481-258
ReadiLink™ Rapid iFluor® 594 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Protein Linking and Labeling Systems
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Red-laser excitable
  • Robust and simple antibody labeling protocol
  • No column purification required
Precaución: For research use only.